The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009179.3 | NP_033205.2 | 4 - 5 | 1910 | 67 |
GenBank mRNA | AK029246.1 | - | 4 - 5 | 1825 | 67 |
AK031052.1 | - | 4 - 5 | 1853 | 67 | |
AK034863.1 | - | 4 - 5 | 1851 | 67 | |
AK053827.1 | - | 4 - 5 | 1849 | 67 | |
AK131653.1 | - | 3 - 4 | 733 | 67 | |
BC015264.1 | - | 3 - 4 | 1152 | 67 | |
BC066064.1 | - | 4 - 5 | 1809 | 67 | |
X76989.1 | - | 3 - 4 | 816 | 67 |