Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stil
Entrez Gene ID
20460
Gene Name
Scl/Tal1 interrupting locus
Gene Aliases
Sil
Chromosome Location
Chr.4: 115000107 - 115043203 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009185.3 NP_033211.2 13 - 14 2633 80
GenBank mRNA AK019471.1 - 2 - 3 986 80
AK049223.1 - 13 - 14 2570 80
AK049550.1 - 12 - 13 1634 80
BC049865.1 - 13 - 14 2587 80
BC060706.1 - 14 - 15 2655 80
BC087545.1 - 12 - 13 2324 80
BC108373.1 - 12 - 13 2301 80
BC141175.1 - 13 - 14 2506 80
BC145493.1 - 13 - 14 2506 80
U36778.1 - 13 - 14 2551 80

View Full Product Details