Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc12a4
Entrez Gene ID
20498
Gene Name
solute carrier family 12, member 4
Gene Aliases
AW546649, KCC1, RBCKCC1
Chromosome Location
Chr.8: 105943590 - 105966149 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253804.1 NP_001240733.1 13 - 14 1825 81
NM_009195.3 NP_033221.1 13 - 14 1819 81
NR_045594.1 13 - 14 1819 81
GenBank mRNA AF047339.1 - 13 - 14 1743 81
AF121118.1 - 13 - 14 1810 81
AK149969.1 - 13 - 14 1808 81
AK151717.1 - 13 - 14 1780 81
AK152762.1 - 13 - 14 1797 81
AK153245.1 - 13 - 14 1797 81
AK159274.1 - 13 - 14 1807 81
AK159331.1 - 13 - 14 1802 81
AK159483.1 - 13 - 14 1775 81
AK159537.1 - 13 - 14 1802 81
AK160034.1 - 13 - 14 1768 81
AK160072.1 - 13 - 14 1802 81
AK164479.1 - 13 - 14 1784 81
AK165874.1 - 13 - 14 1797 81
AK166304.1 - 13 - 14 1778 81
AK167371.1 - 13 - 14 1816 81
AK167444.1 - 13 - 14 1816 81
AK169044.1 - 13 - 14 1774 81
BC066872.1 - 13 - 14 1749 81

View Full Product Details