The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009302.3 | NP_033328.3 | 1 - 2 | 205 | 58 |
GenBank mRNA | AF053974.1 | - | 1 - 2 | 97 | 58 |
AK019882.1 | - | 1 - 2 | 148 | 58 | |
AK041505.1 | - | 1 - 2 | 153 | 58 | |
AK045295.1 | - | 1 - 2 | 177 | 58 | |
AK146773.1 | - | 1 - 2 | 197 | 58 | |
AK172990.1 | - | 1 - 2 | 142 | 58 | |
BC065136.1 | - | 1 - 2 | 147 | 58 | |
BY118458.1 | - | 1 - 2 | 206 | 58 |