The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009864.2 | NP_033994.1 | 3 - 4 | 520 | 84 |
GenBank mRNA | AK076369.1 | - | 3 - 4 | 521 | 84 |
AK166267.1 | - | 3 - 4 | 521 | 84 | |
BC098501.1 | - | 3 - 4 | 509 | 84 | |
X06115.1 | - | 3 - 4 | 461 | 84 |