The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198839.1 | NP_001185768.1 | 4 - 5 | 1015 | 112 |
NM_009954.3 | NP_034084.2 | 4 - 5 | 1078 | 112 | |
GenBank mRNA | AK090315.1 | - | 4 - 5 | 1085 | 112 |
AK145863.1 | - | 4 - 5 | 1079 | 112 | |
AK159168.1 | - | 4 - 5 | 982 | 112 | |
AK166248.1 | - | 4 - 5 | 953 | 112 | |
AK167352.1 | - | 4 - 5 | 1016 | 112 | |
AK167504.1 | - | 4 - 5 | 1016 | 112 | |
AK167516.1 | - | 4 - 5 | 1079 | 112 | |
AK167717.1 | - | 4 - 5 | 1079 | 112 | |
BC057578.1 | - | 4 - 5 | 1038 | 112 | |
U28151.1 | - | 4 - 5 | 989 | 112 | |
U48853.1 | - | 4 - 5 | 973 | 112 |