The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010176.4 | NP_034306.2 | 1 - 2 | 300 | 65 |
GenBank mRNA | AK143759.1 | - | 1 - 2 | 129 | 65 |
AK158808.1 | - | 1 - 2 | 186 | 65 | |
BC010767.1 | - | 1 - 2 | 85 | 65 | |
BY293890.1 | - | 1 - 2 | 282 | 65 | |
M84145.1 | - | 1 - 2 | 126 | 65 | |
Z11774.1 | - | 1 - 2 | 112 | 65 |