The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271467.1 | NP_001258396.1 | 4 - 5 | 604 | 87 |
NM_001271468.1 | NP_001258397.1 | 3 - 4 | 506 | 87 | |
NM_001271470.1 | NP_001258399.1 | 4 - 5 | 671 | 87 | |
NM_001271492.1 | NP_001258421.1 | 3 - 4 | 506 | 87 | |
NM_010249.2 | NP_034379.1 | 3 - 4 | 506 | 87 | |
NM_207669.2 | NP_997552.1 | 3 - 4 | 506 | 87 | |
GenBank mRNA | AK040722.1 | - | 3 - 4 | 507 | 87 |
AK052828.1 | - | - | 482 | 87 | |
AK139710.1 | - | 4 - 5 | 671 | 87 | |
AK140222.1 | - | 4 - 5 | 557 | 87 | |
AK140907.1 | - | 3 - 4 | 417 | 87 | |
AK146373.1 | - | 3 - 4 | 370 | 87 | |
AY282802.1 | - | 1 - 2 | 158 | 87 | |
AY282805.1 | - | 1 - 2 | 158 | 87 | |
BC013558.1 | - | 3 - 4 | 397 | 87 | |
CF730595.1 | - | 3 - 4 | 359 | 87 | |
M74516.1 | - | 3 - 4 | 401 | 87 | |
M74517.1 | - | 3 - 4 | 400 | 87 |