Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gabpb1
Entrez Gene ID
14391
Gene Name
GA repeat binding protein, beta 1
Gene Aliases
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4Tf1B, GABPB, GABPB-1, GABPB-2, GABPB1-1, GABPB1-2, NRF2B1, NRF2B2
Chromosome Location
Chr.2: 126628907 - 126676677 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271468.1 NP_001258397.1 6 - 7 933 61
NM_010249.2 NP_034379.1 6 - 7 933 61
NR_073183.1 4 - 5 626 61
GenBank mRNA AK052828.1 - - 909 61
AK146373.1 - 6 - 7 797 61
AY282800.1 - 3 - 4 390 61
AY282802.1 - 4 - 5 585 61
AY282803.1 - 3 - 4 390 61
AY282804.1 - 4 - 5 472 61
AY282805.1 - 3 - 4 473 61
BC013558.1 - 6 - 7 824 61
CA552206.1 - 3 - 4 350 61
M74517.1 - 6 - 7 827 61

View Full Product Details