The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010752.3 | NP_034882.2 | 16 - 17 | 1865 | 66 |
GenBank mRNA | AF083812.1 | - | 16 - 17 | 1872 | 66 |
AF083813.1 | - | 15 - 16 | 1781 | 66 | |
AK136981.1 | - | 16 - 17 | 1868 | 66 | |
AK169976.1 | - | 5 - 6 | 834 | 66 | |
BC010702.1 | - | 16 - 17 | 1863 | 66 |