The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010765.2 | NP_034895.1 | 1 - 2 | 728 | 81 |
GenBank mRNA | AF039840.1 | - | 1 - 2 | 742 | 81 |
AY533679.1 | - | 1 - 2 | 36 | 81 | |
AY533680.2 | - | 1 - 2 | 36 | 81 | |
BC019184.1 | - | 1 - 2 | 150 | 81 | |
BY722898.1 | - | 1 - 2 | 313 | 81 |