Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Msh3
Entrez Gene ID
17686
Gene Name
mutS homolog 3 (E. coli)
Gene Aliases
D13Em1, Rep-3, Rep3
Chromosome Location
Chr.13: 92211879 - 92355003 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010829.2 NP_034959.2 2 - 3 362 68
GenBank mRNA AK011426.1 - 2 - 3 346 68
AK078467.1 - 2 - 3 313 68
AK133907.1 - 2 - 3 355 68
AK161089.1 - 2 - 3 356 68
BC150759.1 - 2 - 3 281 68
BY710130.1 - 2 - 3 344 68
M24919.1 - 2 - 3 645 68
M80360.1 - 2 - 3 359 68

View Full Product Details