The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011284.3 | NP_035414.3 | 6 - 7 | 623 | 77 |
GenBank mRNA | AK078006.1 | - | 6 - 7 | 624 | 77 |
AK168680.1 | - | 6 - 7 | 623 | 77 | |
AK169847.1 | - | 6 - 7 | 625 | 77 | |
D00812.1 | - | 6 - 7 | 595 | 77 |