Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Shcbp1
Entrez Gene ID
20419
Gene Name
Shc SH2-domain binding protein 1
Gene Aliases
mPAL
Chromosome Location
Chr.8: 4735980 - 4779534 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011369.2 NP_035499.1 2 - 3 282 127
GenBank mRNA AF017152.1 - 2 - 3 282 127
AK137496.1 - 2 - 3 298 127
AK144966.1 - 2 - 3 305 127
AK149580.1 - 2 - 3 299 127
AK162943.1 - 2 - 3 296 127
BC070455.1 - 2 - 3 302 127

View Full Product Details