Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shcbp1
Entrez Gene ID
20419
Gene Name
Shc SH2-domain binding protein 1
Gene Aliases
mPAL
Chromosome Location
Chr.8: 4735980 - 4779534 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011369.2 NP_035499.1 3 - 4 400 73
GenBank mRNA AF017152.1 - 3 - 4 400 73
AK052944.1 - 2 - 3 549 73
AK137496.1 - 3 - 4 416 73
AK144966.1 - 3 - 4 423 73
AK149580.1 - 3 - 4 417 73
AK162943.1 - 3 - 4 414 73
BC070455.1 - 3 - 4 420 73

View Full Product Details