Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St6galnac1
Entrez Gene ID
20445
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1
Gene Aliases
Siat7a
Chromosome Location
Chr.11: 116765025 - 116775507 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011371.2 NP_035501.2 2 - 3 718 84
GenBank mRNA BC141095.1 - - 701 84
BC145373.1 - - 701 84
BC145374.1 - 2 - 3 701 84
Y11274.1 - 2 - 3 718 84

View Full Product Details