The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011371.2 | NP_035501.2 | 7 - 8 | 1396 | 62 |
GenBank mRNA | AK155119.1 | - | - | 576 | 62 |
BC141095.1 | - | - | 1379 | 62 | |
BC145373.1 | - | - | 1379 | 62 | |
BC145374.1 | - | 7 - 8 | 1346 | 62 | |
BE847090.1 | - | 1 - 2 | 60 | 62 | |
Y11274.1 | - | 7 - 8 | 1396 | 62 |