Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smn1
Entrez Gene ID
20595
Gene Name
survival motor neuron 1
Gene Aliases
AI849087, Gemin1, Smn
Chromosome Location
Chr.13: 100123205 - 100137699 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252629.1 NP_001239558.1 4 - 5 567 79
NM_011420.2 NP_035550.1 4 - 5 503 79
GenBank mRNA AK144778.1 - 4 - 5 504 79
AK152495.1 - 4 - 5 477 79
AK167832.1 - 4 - 5 501 79
AK214738.1 - 1 - 2 133 79
BC045158.1 - 4 - 5 467 79
EU234021.1 - 4 - 5 567 79
U63294.1 - 4 - 5 481 79
U77714.1 - 4 - 5 464 79
Y12835.1 - 4 - 5 489 79

View Full Product Details