The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011477.3 | NP_035607.2 | 2 - 2 | 97 | 73 |
GenBank mRNA | AY158995.1 | - | 2 - 2 | 73 | |
BC116866.1 | - | 2 - 2 | 73 | ||
BC116868.1 | - | 2 - 2 | 73 |