The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109905.2 | NP_001103375.1 | 2 - 3 | 407 | 59 |
NM_001109906.2 | NP_001103376.1 | 2 - 3 | 407 | 59 | |
NM_001291160.1 | NP_001278089.1 | 2 - 3 | 407 | 59 | |
NM_011490.4 | NP_035620.1 | 2 - 3 | 407 | 59 | |
NR_111895.1 | 1 - 2 | 141 | 59 | ||
GenBank mRNA | AF061942.1 | - | 2 - 3 | 407 | 59 |
AF395842.1 | - | 1 - 2 | 128 | 59 | |
AK005002.2 | - | 2 - 3 | 348 | 59 | |
AK139532.1 | - | 2 - 3 | 354 | 59 | |
AK145826.1 | - | 2 - 3 | 374 | 59 | |
AK151835.1 | - | 2 - 3 | 213 | 59 | |
AK152269.1 | - | 2 - 3 | 219 | 59 | |
AK152744.1 | - | 2 - 3 | 213 | 59 | |
AK159602.1 | - | 2 - 3 | 324 | 59 | |
AK159781.1 | - | 2 - 3 | 377 | 59 | |
AK160979.1 | - | 2 - 3 | 348 | 59 | |
AK218209.1 | - | 2 - 3 | 168 | 59 | |
AY391773.1 | - | 1 - 2 | 140 | 59 | |
BC012959.1 | - | 2 - 3 | 371 | 59 | |
BC082277.1 | - | 2 - 3 | 344 | 59 |