Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fto
Entrez Gene ID
26383
Gene Name
fat mass and obesity associated
Gene Aliases
AW743446, mKIAA1752
Chromosome Location
Chr.8: 91313491 - 91668433 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011936.2 NP_036066.2 8 - 9 1404 89
GenBank mRNA AJ237917.1 - 8 - 9 1390 89
AK016860.1 - 5 - 6 646 89
AK040866.1 - 8 - 9 1382 89
AK045465.1 - 8 - 9 1381 89
AK049502.1 - 8 - 9 1397 89
AK088881.1 - 8 - 9 1390 89
BC022222.1 - 8 - 9 1355 89
BC057008.1 - 8 - 9 1370 89

View Full Product Details