Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
St6galnac5
Entrez Gene ID
26938
Gene Name
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
Gene Aliases
AI851940, Siat7e
Chromosome Location
Chr.3: 152820709 - 152982207 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012028.4 NP_036158.3 3 - 4 859 63
GenBank mRNA AB028840.1 - - 841 63
AB030836.1 - 3 - 4 862 63
AK034387.1 - - 863 63
AK038434.1 - 3 - 4 863 63
AK042683.1 - 3 - 4 899 63
BC055737.1 - 3 - 4 859 63

View Full Product Details