The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013664.3 | NP_038692.1 | 7 - 8 | 909 | 107 |
GenBank mRNA | AK154836.1 | - | 7 - 8 | 908 | 107 |
AK162715.1 | - | 7 - 8 | 837 | 107 | |
AK167505.1 | - | 7 - 8 | 851 | 107 | |
BC138602.1 | - | 7 - 8 | 848 | 107 | |
BC138603.1 | - | 7 - 8 | 848 | 107 | |
U58885.1 | - | 7 - 8 | 848 | 107 |