Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clnk
Entrez Gene ID
27278
Gene Name
cytokine-dependent hematopoietic cell linker
Gene Aliases
MIST
Chromosome Location
Chr.5: 38705588 - 38876858 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013748.3 NP_038776.3 10 - 11 627 145
GenBank mRNA AB021220.1 - 10 - 11 747 145
AF187819.1 - 10 - 11 627 145
AK082826.1 - 6 - 7 365 145
BC125637.1 - 10 - 11 624 145
BC125639.1 - 10 - 11 624 145

View Full Product Details