Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sytl4
Entrez Gene ID
27359
Gene Name
synaptotagmin-like 4
Gene Aliases
Gph, Slp4
Chromosome Location
Chr.X: 133936385 - 133981822 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290717.1 NP_001277646.1 11 - 12 1401 75
NM_001290718.1 NP_001277647.1 11 - 12 1374 75
NM_001290719.1 NP_001277648.1 11 - 12 1401 75
NM_013757.2 NP_038785.1 11 - 12 1401 75
GenBank mRNA AB025258.1 - 11 - 12 1372 75
AB025259.1 - 11 - 12 1372 75
AB120510.1 - 10 - 11 1187 75
AK030401.1 - 11 - 12 1248 75
AK161582.1 - 11 - 12 1367 75
BC026819.1 - 11 - 12 1401 75

View Full Product Details