The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160404.1 | NP_001153876.1 | 3 - 4 | 383 | 66 |
NM_013814.3 | NP_038842.3 | 3 - 4 | 471 | 66 | |
GenBank mRNA | AK083391.1 | - | 3 - 4 | 384 | 66 |
BC056215.1 | - | 3 - 4 | 388 | 66 | |
BC090962.1 | - | 2 - 3 | 261 | 66 | |
BY711053.1 | - | 3 - 4 | 433 | 66 | |
U73820.1 | - | 1 - 2 | 136 | 66 |