The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013873.3 | NP_038901.3 | 5 - 6 | 756 | 68 |
GenBank mRNA | AF059257.1 | - | 5 - 6 | 632 | 68 |
AK003034.1 | - | 5 - 6 | 617 | 68 | |
AK010293.1 | - | 5 - 6 | 612 | 68 | |
AK032195.1 | - | 5 - 6 | 642 | 68 | |
AK046368.1 | - | 5 - 6 | 635 | 68 | |
AK159034.1 | - | 5 - 6 | 648 | 68 | |
BC051132.1 | - | 5 - 6 | 629 | 68 | |
BC054757.1 | - | 5 - 6 | 756 | 68 |