Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxl6
Entrez Gene ID
30840
Gene Name
F-box and leucine-rich repeat protein 6
Gene Aliases
AU021795, Fbl6
Chromosome Location
Chr.15: 76535723 - 76538746 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013909.2 NP_038937.2 6 - 7 1002 132
GenBank mRNA AB041571.1 - 5 - 6 1240 132
AF176522.1 - 6 - 7 990 132
AK040458.1 - 4 - 5 1382 132
AK052585.1 - 3 - 4 1415 132
BC011485.1 - 4 - 5 1375 132
BC026818.1 - 4 - 5 1369 132
BC032263.1 - 3 - 4 132
BC044784.2 - 5 - 6 1202 132
BC054377.1 - 5 - 6 823 132

View Full Product Details