Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxl12
Entrez Gene ID
30843
Gene Name
F-box and leucine-rich repeat protein 12
Gene Aliases
3110048D16Rik, Fbl12
Chromosome Location
Chr.9: 20637749 - 20644767 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013911.3 NP_038939.2 1 - 2 603 95
GenBank mRNA AF176525.1 - 1 - 2 485 95
AF313405.1 - 1 - 2 91 95
AK012811.1 - 1 - 2 98 95
AK137282.1 - 2 - 3 2270 95
BC054471.1 - 1 - 2 603 95

View Full Product Details