Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galnt3
Entrez Gene ID
14425
Gene Name
UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3
Gene Aliases
-
Chromosome Location
Chr.2: 66082766 - 66124793 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015736.2 NP_056551.2 6 - 7 1571 93
GenBank mRNA AK019995.1 - 6 - 7 1570 93
AK135489.1 - 6 - 7 1532 93
BC043331.1 - 6 - 7 1427 93
U70538.1 - 6 - 7 1415 93

View Full Product Details