The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016676.5 | NP_057885.1 | 1 - 2 | 677 | 105 |
GenBank mRNA | AB232604.1 | - | 1 - 2 | 126 | 105 |
AF035646.1 | - | 1 - 2 | 367 | 105 | |
AK008725.1 | - | 1 - 2 | 667 | 105 | |
AK028320.1 | - | 1 - 2 | 648 | 105 | |
AK087964.1 | - | 1 - 2 | 639 | 105 | |
AK134988.1 | - | 1 - 2 | 597 | 105 | |
AK146357.1 | - | 1 - 2 | 678 | 105 | |
AK150319.1 | - | 1 - 2 | 637 | 105 | |
AK150384.1 | - | 1 - 2 | 331 | 105 | |
AK150791.1 | - | 1 - 2 | 627 | 105 | |
AK151473.1 | - | 1 - 2 | 331 | 105 | |
AK153315.1 | - | 1 - 2 | 637 | 105 | |
AK153325.1 | - | 1 - 2 | 627 | 105 | |
AK159383.1 | - | 1 - 2 | 642 | 105 | |
AK159845.1 | - | 1 - 2 | 642 | 105 | |
AK159988.1 | - | 1 - 2 | 636 | 105 | |
AK160960.1 | - | 1 - 2 | 632 | 105 | |
AK166010.1 | - | 1 - 2 | 648 | 105 | |
AK168320.1 | - | 1 - 2 | 637 | 105 | |
AK195457.1 | - | 1 - 2 | 113 | 105 | |
BC052735.1 | - | - | 627 | 105 | |
BC056374.1 | - | 1 - 2 | 628 | 105 | |
CT010394.1 | - | 1 - 2 | 126 | 105 |