The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016737.2 | NP_058017.1 | 12 - 13 | 1523 | 84 |
GenBank mRNA | AK075988.1 | - | 12 - 13 | 1458 | 84 |
AK088494.1 | - | 12 - 13 | 1465 | 84 | |
AK149493.1 | - | 12 - 13 | 1439 | 84 | |
AK161645.1 | - | 12 - 13 | 1442 | 84 | |
AK167273.1 | - | 12 - 13 | 1457 | 84 | |
AK168181.1 | - | 12 - 13 | 1439 | 84 | |
AK178711.1 | - | 4 - 5 | 358 | 84 | |
BC003794.1 | - | 12 - 13 | 1432 | 84 | |
U27830.1 | - | 12 - 13 | 1440 | 84 |