The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252347.1 | NP_001239276.1 | 1 - 2 | 382 | 61 |
NM_016759.4 | NP_058039.1 | 1 - 2 | 382 | 61 | |
GenBank mRNA | BC046319.1 | - | 1 - 2 | 282 | 61 |
BY258909.1 | - | 1 - 2 | 355 | 61 | |
U73941.1 | - | 1 - 2 | 109 | 61 |