The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164411.1 | NP_001157883.1 | 4 - 5 | 789 | 124 |
NM_001164412.1 | NP_001157884.1 | 4 - 5 | 789 | 124 | |
NM_001164413.1 | NP_001157885.1 | 4 - 5 | 789 | 124 | |
NM_001164414.1 | NP_001157886.1 | 4 - 5 | 789 | 124 | |
NM_016877.4 | NP_058573.3 | 4 - 5 | 789 | 124 | |
GenBank mRNA | AK029753.1 | - | 4 - 5 | 757 | 124 |
AK032248.1 | - | 4 - 5 | 769 | 124 | |
AK075887.1 | - | 4 - 5 | 777 | 125 | |
AK144760.1 | - | 4 - 5 | 747 | 124 | |
AK165997.1 | - | 4 - 5 | 775 | 124 | |
BC048243.1 | - | 4 - 5 | 677 | 124 | |
BC058778.1 | - | 4 - 5 | 741 | 124 | |
U71269.1 | - | 4 - 5 | 775 | 124 |