The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016917.2 | NP_058613.2 | 3 - 4 | 601 | 75 |
GenBank mRNA | AF215637.1 | - | 3 - 4 | 554 | 75 |
AF226613.1 | - | 3 - 4 | 567 | 75 | |
AF231120.1 | - | 3 - 4 | 540 | 75 | |
AK008700.1 | - | 3 - 4 | 405 | 75 | |
AK033987.1 | - | 3 - 4 | 617 | 75 | |
AK144780.1 | - | 3 - 4 | 588 | 75 | |
AK147137.1 | - | 3 - 4 | 587 | 75 | |
AK159294.1 | - | 3 - 4 | 590 | 75 | |
AK159855.1 | - | 3 - 4 | 587 | 75 | |
AK167608.1 | - | 3 - 4 | 590 | 75 | |
BC003438.1 | - | 3 - 4 | 570 | 75 |