Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sart3
Entrez Gene ID
53890
Gene Name
squamous cell carcinoma antigen recognized by T cells 3
Gene Aliases
AU045857, mKIAA0156
Chromosome Location
Chr.5: 113742444 - 113772505 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016926.1 NP_058622.1 12 - 13 1578 66
GenBank mRNA AF172722.1 - 12 - 13 1578 66
AK053828.1 - 12 - 13 1582 66
AK086398.1 - 12 - 13 1579 66
AK129067.1 - 12 - 13 1573 66
BC036350.1 - 12 - 13 1536 66
BC057156.1 - 12 - 13 1569 66

View Full Product Details