Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctnnal1
Entrez Gene ID
54366
Gene Name
catenin (cadherin associated protein), alpha-like 1
Gene Aliases
ACRP, AI616177, AW545119, C86009, Catnal1
Chromosome Location
Chr.4: 56810935 - 56865211 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018761.3 NP_061231.3 2 - 3 424 79
GenBank mRNA AF006071.1 - 2 - 3 411 79
AK034393.1 - 2 - 3 381 79
AK082755.1 - 2 - 3 367 79
BC050070.1 - 2 - 3 401 79
CF896873.1 - 2 - 3 351 79

View Full Product Details