The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081249.1 | NP_001074718.1 | 13 - 14 | 10205 | 90 |
NM_001134474.1 | NP_001127946.1 | 12 - 13 | 4988 | 90 | |
NM_001134475.1 | NP_001127947.1 | 11 - 12 | 2108 | 90 | |
NM_019389.2 | NP_062262.2 | 12 - 13 | 7325 | 90 | |
GenBank mRNA | AK045314.1 | - | 6 - 7 | 3660 | 90 |
AK161227.1 | - | 1 - 2 | 150 | 90 | |
D16263.1 | - | 11 - 12 | 6897 | 90 | |
D28599.1 | - | 13 - 14 | 9949 | 90 | |
D32040.1 | - | 11 - 12 | 1840 | 90 |