The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019710.2 | NP_062684.2 | 15 - 16 | 2530 | 65 |
GenBank mRNA | AF047600.1 | - | 15 - 16 | 2530 | 65 |
BC131667.1 | - | 15 - 16 | 2483 | 65 |