Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cyp11a1
Entrez Gene ID
13070
Gene Name
cytochrome P450, family 11, subfamily a, polypeptide 1
Gene Aliases
Cyp11a, Cypxia1, D9Ertd411e, P450scc, Scc, cscc
Chromosome Location
Chr.9: 58015017 - 58027023 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019779.3 NP_062753.3 3 - 4 640 66
GenBank mRNA AF195119.1 - 3 - 4 640 66
BC068264.1 - 3 - 4 659 66

View Full Product Details