Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Copz1
Entrez Gene ID
56447
Gene Name
coatomer protein complex, subunit zeta 1
Gene Aliases
5930435A22Rik, AA407760, D4Ertd360e
Chromosome Location
Chr.15: 103272834 - 103299865 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019817.1 NP_062791.1 5 - 6 320 79
GenBank mRNA AB037370.1 - 5 - 6 320 79
AK003302.1 - 5 - 6 328 79
AK083824.1 - 5 - 6 340 79
AK156758.1 - 5 - 6 339 79
AK167636.1 - 5 - 6 328 79
AK168382.1 - 5 - 6 339 79
AK168471.1 - 5 - 6 339 79
BC002246.1 - 5 - 6 326 79
BC023207.1 - 4 - 5 304 79
BC025041.1 - 5 - 6 326 79
BC039920.1 - 5 - 6 349 79
BC052891.1 - 5 - 6 331 79
BC058524.1 - 5 - 6 319 79
BC062203.1 - 5 - 6 336 79
BC085314.1 - 5 - 6 341 79

View Full Product Details