Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctsf
Entrez Gene ID
56464
Gene Name
cathepsin F
Gene Aliases
AI481912
Chromosome Location
Chr.19: 4855129 - 4860912 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019861.1 NP_063914.1 8 - 9 1040 61
GenBank mRNA AF136280.1 - 8 - 9 941 61
AF197480.1 - 8 - 9 985 61
AJ131851.1 - 8 - 9 917 61
AK075862.1 - 8 - 9 933 61
AK207816.1 - 1 - 2 59 61
BC004054.1 - 5 - 6 511 61
BC051131.1 - 7 - 8 1159 61
BC058758.1 - 8 - 9 931 61
BC115438.1 - 2 - 3 160 61

View Full Product Details