Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctsf
Entrez Gene ID
56464
Gene Name
cathepsin F
Gene Aliases
AI481912
Chromosome Location
Chr.19: 4855129 - 4860912 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019861.1 NP_063914.1 11 - 12 1298 73
GenBank mRNA AF136280.1 - 11 - 12 1199 73
AF197480.1 - 11 - 12 1243 73
AJ131851.1 - 11 - 12 1175 73
AK075862.1 - 11 - 12 1191 73
AK181867.1 - 1 - 2 74 74
BC004054.1 - 8 - 9 769 73
BC051131.1 - 10 - 11 1417 73
BC058758.1 - 11 - 12 1189 73
BC115438.1 - 5 - 6 418 73

View Full Product Details