The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164733.1 | NP_001158205.1 | 11 - 12 | 1434 | 141 |
NM_001164734.1 | NP_001158206.1 | 11 - 12 | 1444 | 141 | |
NM_019939.2 | NP_064323.2 | 10 - 11 | 1402 | 141 | |
GenBank mRNA | AF161181.1 | - | 10 - 11 | 1266 | 141 |
AF199009.1 | - | 10 - 11 | 1398 | 141 | |
AF199010.1 | - | 11 - 12 | 1440 | 141 | |
AK144434.1 | - | 11 - 12 | 1445 | 141 | |
AK187690.1 | - | 2 - 3 | 141 | ||
BC138666.1 | - | 10 - 11 | 1260 | 141 | |
BC145363.1 | - | 11 - 12 | 1302 | 141 | |
BC145364.1 | - | 11 - 12 | 1292 | 141 |