The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020565.2 | NP_065590.2 | 6 - 7 | 824 | 94 |
GenBank mRNA | AF026075.1 | - | 6 - 7 | 94 | |
BC125593.1 | - | 6 - 7 | 824 | 94 | |
BC125595.1 | - | 6 - 7 | 824 | 94 |