The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020568.3 | NP_065593.2 | 4 - 5 | 376 | 62 |
GenBank mRNA | AF064748.1 | - | 4 - 5 | 387 | 62 |
AK173286.1 | - | 4 - 5 | 296 | 62 |