Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnf25
Entrez Gene ID
57751
Gene Name
ring finger protein 25
Gene Aliases
0610009H16Rik, AO7
Chromosome Location
Chr.1: 74593748 - 74601431 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF171060.1 - 4 - 5 301 68
AK002399.1 - 4 - 5 322 68
AK171752.1 - 4 - 5 299 68
BC022715.1 - 4 - 5 287 68
BC037100.1 - 4 - 5 312 68
BC048821.1 - 4 - 5 309 68
BG081026.2 - 4 - 5 268 68

View Full Product Details