Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdon
Entrez Gene ID
57810
Gene Name
cell adhesion molecule-related/down-regulated by oncogenes
Gene Aliases
CDO, Cd0, Orcam
Chromosome Location
Chr.9: 35421129 - 35507652 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021339.2 NP_067314.2 4 - 5 665 64
GenBank mRNA AF090866.1 - 4 - 5 665 64
AK040711.1 - 5 - 6 757 64
AK157750.1 - 6 - 7 848 64
BC109176.2 - 4 - 5 664 64

View Full Product Details