The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021339.2 | NP_067314.2 | 13 - 14 | 2670 | 68 |
GenBank mRNA | AF090866.1 | - | 13 - 14 | 2670 | 68 |
BC109176.2 | - | 13 - 14 | 2669 | 68 | |
CV557123.1 | - | 2 - 3 | 283 | 68 |