Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxj2
Entrez Gene ID
60611
Gene Name
forkhead box J2
Gene Aliases
Fhx
Chromosome Location
Chr.6: 122820184 - 122845366 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021899.3 NP_068699.1 6 - 7 1616 100
GenBank mRNA AB221666.1 - 5 - 6 815 100
AF253052.1 - 6 - 7 1628 100
BC040395.1 - 7 - 8 1246 100

View Full Product Details